Assay Design



Gene Transcripts

Gene Symbol
prkaca
Entrez Gene ID
549157
Gene Name
protein kinase, cAMP-dependent, alpha catalytic subunit
Gene Aliases
-
Chromosome Location
Chr.3: 123796664 - 123826410 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016403.2 NP_001016403.1 5 - 6 443 96
XM_012953064.1 XP_012808518.1 5 - 6 831 96
XM_012953065.1 XP_012808519.1 6 - 7 690 96
XM_012953066.2 XP_012808520.1 6 - 7 669 96
XM_012953067.1 XP_012808521.1 6 - 7 625 96
XM_012953069.2 XP_012808523.1 6 - 7 604 96
XM_018092085.1 XP_017947574.1 5 - 6 485 96
GenBank mRNA BC170934.1 - 5 - 6 441 96
BC170936.1 - 5 - 6 441 96
CR761141.2 - 5 - 6 443 96
FN562397.1 - 5 - 6 513 96

View Full Product Details