Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001016426.2 | NP_001016426.1 | 3 - 4 | 393 | 68 |
GenBank mRNA | BC135626.1 | - | 3 - 4 | 429 | 68 |
CR761064.2 | - | 3 - 4 | 393 | 68 | |
CR761065.2 | - | 3 - 4 | 393 | 68 |