Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001016459.2 | NP_001016459.1 | 2 - 3 | 214 | 69 |
XM_012962360.2 | XP_012817814.1 | 2 - 3 | 356 | 69 | |
GenBank mRNA | BC170918.1 | - | 2 - 3 | 205 | 69 |
BC170920.1 | - | 2 - 3 | 205 | 69 | |
CR760957.2 | - | 2 - 3 | 214 | 69 |