Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001016465.2 | NP_001016465.1 | 8 - 9 | 768 | 102 |
XM_012962363.2 | XP_012817817.1 | 8 - 9 | 858 | 102 | |
GenBank mRNA | BC166260.1 | - | 8 - 9 | 759 | 102 |
BC170606.1 | - | 8 - 9 | 768 | 102 | |
BC170985.1 | - | 8 - 9 | 768 | 102 | |
CR760937.2 | - | 8 - 9 | 768 | 102 |