Assay Design



Gene Transcripts

Gene Symbol
tmem241
Entrez Gene ID
549221
Gene Name
transmembrane protein 241
Gene Aliases
c18orf45
Chromosome Location
Chr.6: 93974114 - 93985969 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016467.2 NP_001016467.1 1 - 2 99 101
GenBank mRNA BC170975.1 - 1 - 2 93 101
BC170977.1 - 1 - 2 93 101
CR760933.2 - 1 - 2 99 101
CU025002.1 - 1 - 2 103 101

View Full Product Details