Assay Design



Gene Transcripts

Gene Symbol
tulp1
Entrez Gene ID
549236
Gene Name
tubby like protein 1
Gene Aliases
rp14, tubl1
Chromosome Location
Chr.2: 61261254 - 61372702 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016482.2 NP_001016482.1 10 - 11 1193 70
GenBank mRNA CR760884.2 - 10 - 11 1193 70

View Full Product Details