Assay Design



Gene Transcripts

Gene Symbol
rps13
Entrez Gene ID
549356
Gene Name
ribosomal protein S13
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683404: 740824 - 744165 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016602.2 NP_001016602.1 1 - 2 157 69
GenBank mRNA BC118690.1 - 2 - 3 203 69
CR760551.2 - 1 - 2 157 69

View Full Product Details