Assay Design



Gene Transcripts

Gene Symbol
cat.1
Entrez Gene ID
549470
Gene Name
catalase, gene 1
Gene Aliases
cat
Chromosome Location
Chr.4: 4441151 - 4454971 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016716.2 NP_001016716.1 - 70 105
GenBank mRNA CR760094.2 - - 70 105

View Full Product Details