Assay Design



Gene Transcripts

Gene Symbol
ciao1
Entrez Gene ID
549641
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
wdr39
Chromosome Location
Chr.3: 109159707 - 109165649 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016887.2 NP_001016887.1 6 - 7 868 67
GenBank mRNA BC127279.1 - 6 - 7 862 67
CR848572.2 - 6 - 7 868 67

View Full Product Details