Assay Design



Gene Transcripts

Gene Symbol
dad1
Entrez Gene ID
549694
Gene Name
defender against cell death 1
Gene Aliases
DAD-1, ost2
Chromosome Location
Chr.1: 128925993 - 128931990 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016940.2 NP_001016940.1 2 - 3 379 90
GenBank mRNA BC135130.1 - 2 - 3 394 90
CR761961.2 - 2 - 3 379 90

View Full Product Details