Assay Design



Gene Transcripts

Gene Symbol
dnajc19
Entrez Gene ID
549696
Gene Name
DnaJ heat shock protein family (Hsp40) member C19
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683377: 429446 - 436581 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016942.2 NP_001016942.1 3 - 4 238 91
XM_012964760.2 XP_012820214.1 3 - 4 188 91
GenBank mRNA CR761895.2 - 3 - 4 238 91

View Full Product Details