Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017135.2 | NP_001017135.1 | 5 - 6 | 779 | 105 |
XM_012970449.1 | XP_012825903.1 | 5 - 6 | 827 | 105 | |
XM_012970450.2 | XP_012825904.1 | 5 - 6 | 825 | 105 | |
GenBank mRNA | BC153688.1 | - | 4 - 5 | 749 | 105 |
CR761043.2 | - | 5 - 6 | 779 | 105 |