Assay Design



Gene Transcripts

Gene Symbol
med30
Entrez Gene ID
549995
Gene Name
mediator complex subunit 30
Gene Aliases
-
Chromosome Location
Chr.6: 121778248 - 121781304 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017241.2 NP_001017241.1 2 - 3 351 73
GenBank mRNA CR760181.2 - 2 - 3 351 73

View Full Product Details