Assay Design
Gene Transcripts
- Gene Symbol
- elovl6
- Entrez Gene ID
-
550011
- Gene Name
- ELOVL fatty acid elongase 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 58978459 - 59026309 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CR760034.2 |
- |
2 - 3 |
351 |
77 |
View Full Product Details