Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037250.1 | NP_001032327.1 | 10 - 11 | 2395 | 85 |
XM_012956627.2 | XP_012812081.1 | 11 - 12 | 2521 | 85 | |
XM_012956628.2 | XP_012812082.1 | 10 - 11 | 2462 | 85 | |
XM_012956629.2 | XP_012812083.1 | 11 - 12 | 2518 | 85 | |
XM_012956630.1 | XP_012812084.1 | 10 - 11 | 2475 | 85 | |
XM_012956631.1 | XP_012812085.1 | 10 - 11 | 2475 | 85 | |
XM_012956632.1 | XP_012812086.1 | 10 - 11 | 2475 | 85 | |
XM_012956633.1 | XP_012812087.1 | 10 - 11 | 2475 | 85 | |
XM_012956634.1 | XP_012812088.1 | 10 - 11 | 2475 | 85 | |
XM_018091133.1 | XP_017946622.1 | 11 - 12 | 2592 | 85 | |
XM_018091134.1 | XP_017946623.1 | 10 - 11 | 2418 | 85 | |
GenBank mRNA | BC171045.1 | - | 10 - 11 | 2358 | 85 |
CR759994.2 | - | 10 - 11 | 2395 | 85 |