Assay Design



Gene Transcripts

Gene Symbol
cnppd1
Entrez Gene ID
394532
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
c2orf24, cgi-57
Chromosome Location
Chr.9: 77830454 - 77841475 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203604.2 NP_988935.2 5 - 6 658 73
XM_012970235.2 XP_012825689.1 6 - 7 876 73
GenBank mRNA BC061409.1 - 5 - 6 604 73
CR848649.2 - 5 - 6 658 73

View Full Product Details