Assay Design



Gene Transcripts

Gene Symbol
mrpl53
Entrez Gene ID
448697
Gene Name
mitochondrial ribosomal protein L53
Gene Aliases
-
Chromosome Location
Chr.6: 109055673 - 109061440 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005117.2 NP_001005117.1 1 - 2 165 81
GenBank mRNA BC077056.1 - 1 - 2 119 81
CR926208.2 - 1 - 2 165 81

View Full Product Details