Assay Design



Gene Transcripts

Gene Symbol
casp10
Entrez Gene ID
548432
Gene Name
caspase 10
Gene Aliases
alps2, caspace-10, caspase-10, flice2, mch4, xCaspace-10, xCaspase-10
Chromosome Location
Chr.9: 49158106 - 49194285 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015715.2 NP_001015715.2 6 - 7 910 93
GenBank mRNA BC089646.1 - - 798 93
CR942647.2 - 6 - 7 910 93

View Full Product Details