Assay Design



Gene Transcripts

Gene Symbol
klf2
Entrez Gene ID
549518
Gene Name
Kruppel-like factor 2
Gene Aliases
lklf
Chromosome Location
Chr.1: 100267957 - 100271372 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016764.3 NP_001016764.1 1 - 2 182 102
GenBank mRNA BC122894.1 - 1 - 2 182 102
CR855754.2 - 1 - 2 80 102

View Full Product Details