Assay Design



Gene Transcripts

Gene Symbol
cdc37l1
Entrez Gene ID
733511
Gene Name
cell division cycle 37-like 1
Gene Aliases
cdc37b, harc
Chromosome Location
Chr.1: 115084262 - 115091950 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044440.1 NP_001037905.1 4 - 5 623 109
XM_012964032.2 XP_012819486.1 5 - 6 725 109
XM_012964035.2 XP_012819489.1 5 - 6 712 109
XM_012964039.2 XP_012819493.1 5 - 6 672 109
XM_012964044.2 XP_012819498.1 5 - 6 657 109
GenBank mRNA BC166937.1 - 5 - 6 713 109
CR760680.2 - 4 - 5 623 109

View Full Product Details