Assay Design



Gene Transcripts

Gene Symbol
sult1e1
Entrez Gene ID
733517
Gene Name
sulfotransferase family 1E member 1
Gene Aliases
est-1, ste
Chromosome Location
Chr.4: 38982213 - 38998924 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044443.1 NP_001037908.1 8 - 9 844 69
XM_012961234.1 XP_012816688.1 7 - 8 804 69
XM_012961236.2 XP_012816690.1 7 - 8 796 69
GenBank mRNA BC135544.1 - 8 - 9 842 69
CR760765.2 - 8 - 9 844 69

View Full Product Details