Assay Design



Gene Transcripts

Gene Symbol
parpbp
Entrez Gene ID
733805
Gene Name
PARP1 binding protein
Gene Aliases
PARI, c12orf48
Chromosome Location
Chr.3: 39622086 - 39640007 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045574.1 NP_001039039.1 5 - 6 589 102
GenBank mRNA CR848174.2 - 5 - 6 589 102

View Full Product Details