Assay Design



Gene Transcripts

Gene Symbol
krcc1
Entrez Gene ID
733809
Gene Name
lysine-rich coiled-coil 1
Gene Aliases
chbp2, zmat1, zmat4.2
Chromosome Location
Chr.1: 15173494 - 15194558 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045575.1 NP_001039040.1 6 - 7 924 75
XM_012964252.2 XP_012819706.1 6 - 7 994 75
GenBank mRNA CR848257.2 - 6 - 7 924 75

View Full Product Details