Assay Design



Gene Transcripts

Gene Symbol
polr2j
Entrez Gene ID
733914
Gene Name
polymerase (RNA) II (DNA directed) polypeptide J, 13.3kDa
Gene Aliases
-
Chromosome Location
Chr.2: 90822177 - 90827773 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045630.1 NP_001039095.1 2 - 3 160 104
GenBank mRNA CR942371.2 - 2 - 3 160 104

View Full Product Details