Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045634.1 | NP_001039099.1 | 4 - 5 | 477 | 87 |
XM_012968545.2 | XP_012823999.1 | 4 - 5 | 594 | 87 | |
GenBank mRNA | BC167936.1 | - | 4 - 5 | 758 | 87 |
CR942384.2 | - | 4 - 5 | 477 | 87 |