Assay Design



Gene Transcripts

Gene Symbol
wbp4
Entrez Gene ID
734007
Gene Name
WW domain binding protein 4
Gene Aliases
-
Chromosome Location
Chr.2: 129053175 - 129066249 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045707.1 NP_001039172.1 6 - 7 467 103
XM_012957171.1 XP_012812625.1 6 - 7 534 103
XM_012957173.2 XP_012812627.1 6 - 7 588 103
XM_012957174.2 XP_012812628.1 6 - 7 532 103
XM_012957175.1 XP_012812629.1 6 - 7 565 103
GenBank mRNA BC135946.1 - 6 - 7 471 103
CR761869.2 - 6 - 7 467 103

View Full Product Details