Assay Design



Gene Transcripts

Gene Symbol
wbp4
Entrez Gene ID
734007
Gene Name
WW domain binding protein 4
Gene Aliases
-
Chromosome Location
Chr.2: 129053175 - 129066249 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045707.1 NP_001039172.1 7 - 8 530 78
XM_012957171.1 XP_012812625.1 7 - 8 597 78
XM_012957173.2 XP_012812627.1 7 - 8 651 78
XM_012957174.2 XP_012812628.1 7 - 8 595 78
XM_012957175.1 XP_012812629.1 7 - 8 628 78
GenBank mRNA BC135946.1 - 7 - 8 534 78
CR761869.2 - 7 - 8 530 78

View Full Product Details