Assay Design



Gene Transcripts

Gene Symbol
faf2
Entrez Gene ID
734096
Gene Name
Fas associated factor family member 2
Gene Aliases
etea, faf2-a, faf2-b, ubxd8, ubxn3b
Chromosome Location
Chr.3: 38041219 - 38055933 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045770.1 NP_001039235.1 2 - 3 151 64
GenBank mRNA BC157333.1 - 2 - 3 185 64
CR942713.1 - 2 - 3 151 64

View Full Product Details