Assay Design



Gene Transcripts

Gene Symbol
ralyl
Entrez Gene ID
779659
Gene Name
RALY RNA binding protein like
Gene Aliases
hnrpcl3
Chromosome Location
Chr.6: 109662831 - 109981764 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078745.1 NP_001072213.1 3 - 4 450 81
XM_012964876.2 XP_012820330.1 3 - 4 2474 81
GenBank mRNA BC118782.1 - 3 - 4 450 81

View Full Product Details