Assay Design



Gene Transcripts

Gene Symbol
rabac1
Entrez Gene ID
779672
Gene Name
Rab acceptor 1 (prenylated)
Gene Aliases
pra1, praf1, rabac1-a, rabac1-b, yip3
Chromosome Location
Chr.7: 98546139 - 98551200 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078757.1 NP_001072225.1 3 - 4 1063 67
GenBank mRNA BC118806.1 - 3 - 4 1063 67
BC135390.1 - 4 - 5 442 67

View Full Product Details