Assay Design



Gene Transcripts

Gene Symbol
ehd3
Entrez Gene ID
779678
Gene Name
EH domain containing 3
Gene Aliases
-
Chromosome Location
Chr.5: 3613421 - 3651211 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078763.1 NP_001072231.1 1 - 2 530 68
GenBank mRNA BC118817.1 - 1 - 2 530 68

View Full Product Details