Assay Design



Gene Transcripts

Gene Symbol
ehd3
Entrez Gene ID
779678
Gene Name
EH domain containing 3
Gene Aliases
-
Chromosome Location
Chr.5: 3613421 - 3651211 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078763.1 NP_001072231.1 5 - 6 1649 72
XM_012963135.2 XP_012818589.1 6 - 7 1848 72
GenBank mRNA BC118817.1 - 5 - 6 1649 72

View Full Product Details