Assay Design



Gene Transcripts

Gene Symbol
spata6l
Entrez Gene ID
779679
Gene Name
spermatogenesis associated 6-like
Gene Aliases
c9orf68
Chromosome Location
Chr.1: 115091923 - 115119129 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078764.1 NP_001072232.1 5 - 6 663 95
XM_012964987.2 XP_012820441.1 4 - 5 95
XM_018094302.1 XP_017949791.1 6 - 7 95
XM_018094304.1 XP_017949793.1 5 - 6 95
XM_018094309.1 XP_017949798.1 5 - 6 95
GenBank mRNA BC118818.1 - 5 - 6 663 95
CR760075.2 - 5 - 6 487 95

View Full Product Details