Assay Design



Gene Transcripts

Gene Symbol
smpx
Entrez Gene ID
779706
Gene Name
small muscle protein X-linked
Gene Aliases
DFN6, DFNX4, chisel, csl, zgc:91881
Chromosome Location
Chr.2: 41219935 - 41262560 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078785.1 NP_001072253.1 2 - 3 219 65
GenBank mRNA BC118903.1 - 2 - 3 219 65
CU025181.1 - 2 - 3 208 65

View Full Product Details