Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001078786.1 | NP_001072254.1 | 11 - 12 | 1450 | 68 |
XM_012965024.2 | XP_012820478.1 | 10 - 11 | 1395 | 68 | |
XM_018094323.1 | XP_017949812.1 | 8 - 9 | 802 | 68 | |
GenBank mRNA | BC121219.1 | - | 11 - 12 | 1450 | 68 |