Assay Design



Gene Transcripts

Gene Symbol
tmem259
Entrez Gene ID
779707
Gene Name
transmembrane protein 259
Gene Aliases
c19orf6
Chromosome Location
Chr.1: 81413117 - 81435363 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078786.1 NP_001072254.1 7 - 8 1076 64
XM_012965019.2 XP_012820473.1 7 - 8 1153 64
XM_012965024.2 XP_012820478.1 7 - 8 1154 64
XM_018094323.1 XP_017949812.1 4 - 5 428 64
GenBank mRNA BC121219.1 - 7 - 8 1076 64

View Full Product Details