Assay Design



Gene Transcripts

Gene Symbol
taf4
Entrez Gene ID
779738
Gene Name
TATA-box binding protein associated factor 4
Gene Aliases
taf2c, taf2c1, taf4a, tafii130, tafii135
Chromosome Location
Chr.10: 1088744 - 1118558 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078817.1 NP_001072285.1 6 - 7 1742 80
XM_012952548.2 XP_012808002.1 6 - 7 2311 80
GenBank mRNA BC121308.1 - 6 - 7 1742 80
CR761139.2 - 3 - 4 309 80

View Full Product Details