Assay Design



Gene Transcripts

Gene Symbol
eme1
Entrez Gene ID
779752
Gene Name
essential meiotic structure-specific endonuclease 1
Gene Aliases
-
Chromosome Location
Chr.10: 35314339 - 35319572 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078831.1 NP_001072299.1 - 885 68
GenBank mRNA BC121344.1 - - 885 68
CR855465.2 - - 889 68

View Full Product Details