Assay Design



Gene Transcripts

Gene Symbol
mapk7
Entrez Gene ID
779785
Gene Name
mitogen-activated protein kinase 7
Gene Aliases
bmk1, erk4, erk5, prkm7
Chromosome Location
Chr.9: 2263217 - 2274761 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078864.1 NP_001072332.1 4 - 5 1837 78
XM_012970540.2 XP_012825994.1 4 - 5 1830 78
XM_012970541.2 XP_012825995.1 4 - 5 1667 78
XM_012970542.2 XP_012825996.1 4 - 5 1746 78
GenBank mRNA BC121410.1 - 4 - 5 1837 78

View Full Product Details