Assay Design



Gene Transcripts

Gene Symbol
cdca9
Entrez Gene ID
779847
Gene Name
cell division cycle associated 9
Gene Aliases
Borealin-2, Dasra-A, DasraA, cdca8.2
Chromosome Location
Chr.4: 25159150 - 25170872 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078925.1 NP_001072393.1 2 - 3 336 67
GenBank mRNA BC121590.1 - 2 - 3 336 67

View Full Product Details