Assay Design



Gene Transcripts

Gene Symbol
vopp1
Entrez Gene ID
779880
Gene Name
VOPP1, WBP1/VOPP1 family member
Gene Aliases
ecop, gasp
Chromosome Location
Chr.6: 65597737 - 65665548 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078958.1 NP_001072426.1 4 - 5 662 76
XM_018094698.1 XP_017950187.1 4 - 5 334 76
GenBank mRNA BC121667.1 - 4 - 5 662 76

View Full Product Details