Assay Design



Gene Transcripts

Gene Symbol
gba3
Entrez Gene ID
779892
Gene Name
uncharacterized LOC779892
Gene Aliases
-
Chromosome Location
Chr.1: 22341527 - 22372479 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078970.1 NP_001072438.1 3 - 4 452 88
XM_012965261.2 XP_012820715.1 3 - 4 414 88
XM_018094457.1 XP_017949946.1 3 - 4 440 88
GenBank mRNA BC121688.1 - 3 - 4 452 88

View Full Product Details