Assay Design



Gene Transcripts

Gene Symbol
zbtb44
Entrez Gene ID
779914
Gene Name
zinc finger and BTB domain containing 44
Gene Aliases
btbd15, hspc063
Chromosome Location
Chr.7: 46804798 - 46844655 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078991.1 NP_001072459.1 2 - 3 1150 87
XM_012966742.2 XP_012822196.1 2 - 3 2120 87
GenBank mRNA BC121859.1 - 2 - 3 1150 87

View Full Product Details