Assay Design



Gene Transcripts

Gene Symbol
c1orf21
Entrez Gene ID
780109
Gene Name
chromosome 1 open reading frame 21
Gene Aliases
-
Chromosome Location
Chr.4: 109645733 - 109708595 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079184.1 NP_001072652.1 5 - 6 512 75
XM_012961365.2 XP_012816819.1 5 - 6 544 75
XM_018093103.1 XP_017948592.1 5 - 6 532 75
GenBank mRNA BC123935.1 - 5 - 6 512 75

View Full Product Details