Assay Design



Gene Transcripts

Gene Symbol
art1
Entrez Gene ID
780139
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
art2, cd296, rt6
Chromosome Location
Chr.Un|NW_016683693: 97940 - 101444 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079214.1 NP_001072682.1 1 - 2 849 61
GenBank mRNA BC124013.1 - 1 - 2 849 61

View Full Product Details