Assay Design



Gene Transcripts

Gene Symbol
arrdc2
Entrez Gene ID
780156
Gene Name
arrestin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 80066158 - 80098306 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079231.1 NP_001072699.1 4 - 5 791 76
XM_012965809.2 XP_012821263.1 4 - 5 848 76
GenBank mRNA BC124042.1 - 4 - 5 791 76
BC160577.1 - 4 - 5 737 76

View Full Product Details