Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079343.1 | NP_001072811.1 | 4 - 5 | 377 | 66 |
XM_012964953.1 | XP_012820407.1 | 4 - 5 | 386 | 66 | |
XM_012964954.1 | XP_012820408.1 | 3 - 4 | 304 | 66 | |
GenBank mRNA | BC121487.1 | - | 4 - 5 | 377 | 66 |