Assay Design



Gene Transcripts

Gene Symbol
calb1
Entrez Gene ID
780272
Gene Name
calbindin 1
Gene Aliases
calb, calb1-a, calb1-b, calbindin
Chromosome Location
Chr.6: 111362008 - 111395319 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079343.1 NP_001072811.1 7 - 8 573 84
XM_012964953.1 XP_012820407.1 7 - 8 582 84
XM_012964954.1 XP_012820408.1 6 - 7 500 84
GenBank mRNA BC121487.1 - 7 - 8 573 84

View Full Product Details