Assay Design



Gene Transcripts

Gene Symbol
atp6v1g2
Entrez Gene ID
780301
Gene Name
ATPase H+ transporting V1 subunit G2
Gene Aliases
atp6g, atp6g2, ng38, vma10
Chromosome Location
Chr.8: 53723701 - 53728389 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079372.1 NP_001072840.1 1 - 2 228 67
GenBank mRNA BC121968.1 - 1 - 2 228 67
BC135509.1 - 1 - 2 183 67

View Full Product Details