Assay Design



Gene Transcripts

Gene Symbol
tma16
Entrez Gene ID
549385
Gene Name
translation machinery associated 16 homolog
Gene Aliases
c4orf43
Chromosome Location
Chr.1: 42388191 - 42423317 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016631.3 NP_001016631.1 2 - 3 208 72
XM_012962584.2 XP_012818038.1 3 - 4 523 72
GenBank mRNA BC135799.1 - 2 - 3 208 72
CR760447.2 - 2 - 3 115 72

View Full Product Details